Assay Details
| Gene Symbol: Ampd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276301.1 | NP_001263230.1 | 4 - 5 | 769 | 110 | |
| NM_009667.3 | NP_033797.2 | 4 - 5 | 888 | 110 | ||
| GenBank mRNA | AK133465.1 | - | 4 - 5 | 755 | 110 | |
| AK171276.1 | - | 4 - 5 | 705 | 110 | ||
| BC040366.1 | - | 4 - 5 | 888 | 4503933 | 110 | |
| BC056380.1 | - | 4 - 5 | 730 | 6851948 | 110 | |
| D85596.1 | - | 4 - 5 | 767 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ampd3
Genomic Map