Assay Details
| Gene Symbol: Arhgap17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122643.1 | NP_001116115.1 | 18 - 19 | 2280 | 128 | |
| GenBank mRNA | AK046411.1 | - | 18 - 19 | 2422 | 128 | |
| AK149986.1 | - | 18 - 19 | 2252 | 128 | ||
| BC003259.1 | - | 18 - 19 | 2219 | 3500822 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap17
Genomic Map