Assay Details
| Gene Symbol: Xpo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028816.2 | NP_083092.2 | 10 - 11 | 1954 | 74 | |
| GenBank mRNA | AK049009.1 | - | 10 - 11 | 1967 | 74 | |
| BC058090.1 | - | 10 - 11 | 1954 | 6826356 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xpo6
Genomic Map