Assay Details
| Gene Symbol: Il4ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008700.3 | NP_001008700.1 | 9 - 10 | 1097 | 62 | |
| GenBank mRNA | AF000304.1 | - | 7 - 8 | 874 | 62 | |
| AK034466.1 | - | 9 - 10 | 1141 | 62 | ||
| AK088086.1 | - | 9 - 10 | 1109 | 62 | ||
| AK137827.1 | - | 9 - 10 | 1109 | 62 | ||
| AK150584.1 | - | 9 - 10 | 1109 | 62 | ||
| AK151998.1 | - | 9 - 10 | 1109 | 62 | ||
| AK153186.1 | - | 9 - 10 | 1109 | 62 | ||
| AK159375.1 | - | 9 - 10 | 1108 | 62 | ||
| AK160016.1 | - | 9 - 10 | 1101 | 62 | ||
| BC112911.1 | - | 9 - 10 | 1097 | 30138743 | 62 | |
| BC128301.1 | - | 8 - 9 | 968 | 62 | ||
| BC128302.1 | - | 8 - 9 | 968 | 62 | ||
| KC212685.1 | - | 989 | 62 | |||
| M27959.1 | - | 9 - 10 | 1092 | 62 | ||
| M27960.1 | - | 10 - 11 | 1206 | 62 | ||
| M29854.1 | - | 9 - 10 | 1076 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il4ra
Genomic Map