Assay Details
| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146198.2 | NP_666310.1 | 15 - 16 | 1922 | 83 | |
| GenBank mRNA | AK008219.1 | - | 15 - 16 | 1977 | 83 | |
| BC031742.1 | - | 15 - 16 | 1923 | 4221537 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a11
Genomic Map