Assay Details
| Gene Symbol: Ccdc101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029339.3 | NP_083615.3 | 6 - 7 | 611 | 67 | |
| GenBank mRNA | AK006287.1 | - | 6 - 7 | 521 | 67 | |
| AK139102.1 | - | 6 - 7 | 612 | 67 | ||
| AK169898.1 | - | 6 - 7 | 607 | 67 | ||
| BC026784.1 | - | 6 - 7 | 549 | 4206090 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc101
Genomic Map