Assay Details
| Gene Symbol: Sh2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289538.1 | NP_001276467.1 | 8 - 9 | 2484 | 124 | |
| GenBank mRNA | AF421138.1 | - | 8 - 9 | 2178 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh2b1
Genomic Map