Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700014997.mm.1 |
TC0700001756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014997.mm.1 |
NM_008855.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014997.mm.1 |
NM_008855.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015002.mm.1 |
TC0700001756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015002.mm.1 |
NM_008855.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015002.mm.1 |
NM_008855.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014995.mm.1 |
TC0700001756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014995.mm.1 |
NM_008855.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014995.mm.1 |
NM_008855.2 |
Refseq |