Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019011.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019011.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019011.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019012.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019012.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019012.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019010.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019010.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019010.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019015.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019015.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019015.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019018.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019018.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019018.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019016.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019016.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019016.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019013.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019013.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019013.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019014.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019014.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019014.mm.1 |
TC1700001989.mm.1 |
MTA TC |