Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019020.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019020.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019020.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019019.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019019.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019019.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019022.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019022.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019022.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019028.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019028.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019028.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019029.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019029.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019029.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019024.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019024.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019024.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019021.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019021.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019021.mm.1 |
TC1700001989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019025.mm.1 |
NM_013854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019025.mm.1 |
NM_013854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019025.mm.1 |
TC1700001989.mm.1 |
MTA TC |