Assay Details
| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007504.2 | NP_031530.2 | 2 - 3 | 284 | 93 | |
| GenBank mRNA | AY081946.1 | - | 2 - 3 | 315 | 93 | |
| BC036292.1 | - | 2 - 3 | 284 | 4191741 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp2a1
Genomic Map