Assay Details
| Gene Symbol: Ypel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026875.2 | NP_081151.2 | 1 - 2 | 226 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_026875.2 transcripts.
Set Membership: |
|

Ypel3
Genomic Map