Assay Details
| Gene Symbol: Gdpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024228.2 | NP_077190.2 | 1 - 2 | 200 | 81 | |
| GenBank mRNA | AK003726.1 | - | 1 - 2 | 202 | 81 | |
| BY101331.1 | - | 1 - 2 | 281 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gdpd3
Genomic Map