Assay Details
| Gene Symbol: Gdpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024228.2 | NP_077190.2 | 8 - 9 | 828 | 74 | |
| GenBank mRNA | AK003726.1 | - | 8 - 9 | 830 | 74 | |
| AW045753.1 | - | 2 - 3 | 74 | |||
| BC002172.1 | - | 6 - 7 | 747 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gdpd3
Genomic Map