Assay Details
| Gene Symbol: Setd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178029.3 | NP_821172.2 | 12 - 13 | 3200 | 64 | |
| GenBank mRNA | BC049883.1 | - | 5 - 6 | 588 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Setd1a
Genomic Map