Assay Details
| Gene Symbol: Nkx1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009123.2 | NP_033149.1 | 1 - 2 | 323 | 139 | |
| GenBank mRNA | AF222444.1 | - | 1 - 2 | 323 | 139 | |
| AF222445.1 | - | 1 - 2 | 324 | 139 | ||
| AF223361.1 | - | 1 - 2 | 325 | 139 | ||
| BC139757.1 | - | 1 - 2 | 252 | 40047346 | 139 | |
| X75384.1 | - | 1 - 2 | 428 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nkx1-2
Genomic Map