Assay Details
| Gene Symbol: Htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019564.3 | NP_062510.2 | 2 - 3 | 641 | 65 | |
| GenBank mRNA | AF179369.1 | - | 2 - 3 | 612 | 65 | |
| AK090320.1 | - | 2 - 3 | 356 | 65 | ||
| AK128916.1 | - | 2 - 3 | 614 | 65 | ||
| AK132270.1 | - | 2 - 3 | 614 | 65 | ||
| AK167667.1 | - | 2 - 3 | 207 | 65 | ||
| BC013516.1 | - | 2 - 3 | 611 | 4235912 | 65 | |
| CB193609.1 | - | 2 - 3 | 641 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK167667.1 transcripts.
Set Membership: |
|

Htra1
Genomic Map