Assay Details
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133942.2 | NP_598703.1 | 8 - 9 | 764 | 69 | |
GenBank mRNA | AK044224.1 | - | 10 - 11 | 916 | 69 | |
AK083340.1 | - | 10 - 11 | 938 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
