Assay Details
| Gene Symbol: Tial1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009383.2 | NP_033409.1 | 9 - 10 | 1140 | 123 | |
| GenBank mRNA | AK012085.1 | - | 9 - 10 | 784 | 123 | |
| AK132677.1 | - | 10 - 11 | 1308 | 123 | ||
| BC010496.1 | - | 10 - 11 | 874 | 3497971 | 123 | |
| BC068194.1 | - | 5 - 6 | 416 | 123 | ||
| U55861.1 | - | 9 - 10 | 916 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tial1
Genomic Map