Assay Details
| Gene Symbol: Tial1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009383.2 | NP_033409.1 | 6 - 7 | 858 | 124 | |
| GenBank mRNA | AK012085.1 | - | 6 - 7 | 502 | 124 | |
| AK132677.1 | - | 7 - 8 | 1028 | 124 | ||
| BC010496.1 | - | 7 - 8 | 592 | 3497971 | 124 | |
| BC068194.1 | - | 2 - 3 | 134 | 124 | ||
| U55861.1 | - | 6 - 7 | 634 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tial1
Genomic Map