Assay Details
| Gene Symbol: Tgfb1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289550.1 | NP_001276479.1 | 1 - 2 | 197 | 53 | |
| NM_001289551.1 | NP_001276480.1 | 1 - 2 | 197 | 53 | ||
| NM_001289552.1 | NP_001276481.1 | 1 - 2 | 197 | 53 | ||
| GenBank mRNA | AK003170.1 | - | 1 - 2 | 32 | 53 | |
| AK041233.1 | - | 1 - 2 | 25 | 53 | ||
| AK087114.1 | - | 1 - 2 | 32 | 53 | ||
| AK156423.1 | - | 1 - 2 | 32 | 53 | ||
| AK158409.1 | - | 1 - 2 | 49 | 53 | ||
| BC002049.1 | - | 1 - 2 | 31 | 3489459 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156423.1, AK158409.1, BC002049.1, NM_001289550.1, NM_001289551.1, NM_001289552.1 transcripts.
Set Membership: |
|

Tgfb1i1
Genomic Map