Assay Details
| Gene Symbol: Tgfb1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289550.1 | NP_001276479.1 | 4 - 5 | 510 | 94 | |
| NM_001289551.1 | NP_001276480.1 | 4 - 5 | 510 | 94 | ||
| NM_001289553.1 | NP_001276482.1 | 3 - 4 | 394 | 94 | ||
| GenBank mRNA | AK003170.1 | - | 4 - 5 | 345 | 94 | |
| AK041233.1 | - | 4 - 5 | 338 | 94 | ||
| AK085042.1 | - | 3 - 4 | 236 | 94 | ||
| AK087114.1 | - | 4 - 5 | 345 | 94 | ||
| AK156423.1 | - | 4 - 5 | 345 | 94 | ||
| BC002049.1 | - | 4 - 5 | 344 | 3489459 | 94 | |
| BC056362.1 | - | 3 - 4 | 1619 | 6816930 | 94 | |
| DQ143891.1 | - | 3 - 4 | 395 | 94 | ||
| DQ143892.1 | - | 2 - 3 | 210 | 94 | ||
| DQ143895.1 | - | 2 - 3 | 599 | 94 | ||
| DQ143898.1 | - | 3 - 4 | 472 | 94 | ||
| DQ143899.1 | - | 2 - 3 | 599 | 94 | ||
| DQ143900.1 | - | 3 - 4 | 407 | 94 | ||
| L22482.1 | - | 3 - 4 | 471 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the DQ143892.1, DQ143895.1, DQ143898.1, DQ143899.1, NM_001289553.1 transcripts.
Set Membership: |
|

Tgfb1i1
Genomic Map