Assay Details
| Gene Symbol: Stx4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009294.3 | NP_033320.1 | 9 - 10 | 1037 | 70 | |
| GenBank mRNA | AK161971.1 | - | 9 - 10 | 1030 | 70 | |
| BC005791.1 | - | 9 - 10 | 877 | 3603665 | 70 | |
| BC011491.1 | - | 9 - 10 | 1037 | 4016871 | 70 | |
| BC052023.1 | - | 9 - 10 | 973 | 5711022 | 70 | |
| U76832.1 | - | 9 - 10 | 806 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stx4a
Genomic Map