Assay Details
| Gene Symbol: Caly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190386.1 | NP_001177315.1 | 1 - 2 | 211 | 72 | |
| GenBank mRNA | BC049663.1 | - | 1 - 2 | 178 | 6532801 | 72 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC049663.1, NM_001190386.1 transcripts.
Set Membership: |
|

Caly
Genomic Map