Assay Details
| Gene Symbol: Dpysl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011993.4 | NP_036123.3 | 5 - 6 | 663 | 59 | |
| GenBank mRNA | AB006715.1 | - | 5 - 6 | 592 | 59 | |
| AK090197.1 | - | 5 - 6 | 637 | 59 | ||
| AK145638.1 | - | 4 - 5 | 766 | 59 | ||
| AK165694.1 | - | 5 - 6 | 664 | 59 | ||
| BC046333.1 | - | 4 - 5 | 2216 | 59 | ||
| BC090674.1 | - | 636 | 59 | |||
| BC138573.1 | - | 5 - 6 | 581 | 8861593 | 59 | |
| BC138574.1 | - | 5 - 6 | 581 | 8861594 | 59 | |
| Y09079.1 | - | 5 - 6 | 642 | 59 | ||
More Information
Set Membership: |
|

Dpysl4
Genomic Map