Assay Details
| Gene Symbol: Dpysl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011993.4 | NP_036123.3 | 6 - 7 | 738 | 74 | |
| GenBank mRNA | AB006715.1 | - | 6 - 7 | 667 | 74 | |
| AK145638.1 | - | 5 - 6 | 841 | 74 | ||
| AK165694.1 | - | 6 - 7 | 739 | 74 | ||
| BC046333.1 | - | 5 - 6 | 2291 | 74 | ||
| BC090674.1 | - | 711 | 74 | |||
| BC138573.1 | - | 6 - 7 | 656 | 8861593 | 74 | |
| BC138574.1 | - | 6 - 7 | 656 | 8861594 | 74 | |
| Y09079.1 | - | 6 - 7 | 717 | 74 | ||
More Information
Set Membership: |
|

Dpysl4
Genomic Map