Assay Details
| Gene Symbol: Dpysl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011993.4 | NP_036123.3 | 10 - 11 | 1233 | 81 | |
| GenBank mRNA | AB006715.1 | - | 10 - 11 | 1162 | 81 | |
| AK090197.1 | - | 9 - 10 | 1761 | 81 | ||
| AK145638.1 | - | 9 - 10 | 1336 | 81 | ||
| AK165694.1 | - | 10 - 11 | 1234 | 81 | ||
| BC046333.1 | - | 9 - 10 | 2786 | 81 | ||
| BC090674.1 | - | 1205 | 81 | |||
| BC138573.1 | - | 10 - 11 | 1151 | 8861593 | 81 | |
| BC138574.1 | - | 10 - 11 | 1151 | 8861594 | 81 | |
| Y09079.1 | - | 10 - 11 | 1212 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl4
Genomic Map