Assay Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023059.3 | NP_075546.2 | 6 - 7 | 1221 | 137 | |
| GenBank mRNA | AF239957.1 | - | 5 - 6 | 732 | 137 | |
| AK133518.1 | - | 6 - 7 | 846 | 137 | ||
| AK145889.1 | - | 6 - 7 | 1224 | 137 | ||
| BC010806.1 | - | 5 - 6 | 743 | 4240425 | 137 | |
| BC094069.1 | - | 6 - 7 | 827 | 4974962 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sigirr
Genomic Map