Assay Details
| Gene Symbol: Sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127351.1 | NP_001120823.1 | 2 - 3 | 295 | 102 | |
| NM_001177804.1 | NP_001171275.1 | 2 - 3 | 287 | 102 | ||
| NM_022433.2 | NP_071878.2 | 2 - 3 | 340 | 102 | ||
| GenBank mRNA | AF299338.1 | - | 2 - 3 | 340 | 102 | |
| AF299339.1 | - | 2 - 3 | 295 | 102 | ||
| AK075861.1 | - | 2 - 3 | 269 | 102 | ||
| BC025878.1 | - | 2 - 3 | 239 | 5253854 | 102 | |
| CB953804.1 | - | 2 - 3 | 287 | 102 | ||
| EU886466.1 | - | 2 - 3 | 285 | 102 | ||
| FJ621493.1 | - | 2 - 3 | 283 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF299338.1, AF299339.1, AK075861.1, BC025878.1, NM_001127351.1, NM_022433.2 transcripts.
Set Membership: |
|

Sirt3
Genomic Map