Assay Details
| Gene Symbol: Sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127351.1 | NP_001120823.1 | 5 - 6 | 797 | 85 | |
| NM_001177804.1 | NP_001171275.1 | 5 - 6 | 789 | 85 | ||
| NM_022433.2 | NP_071878.2 | 5 - 6 | 842 | 85 | ||
| GenBank mRNA | AF299338.1 | - | 5 - 6 | 842 | 85 | |
| AF299339.1 | - | 5 - 6 | 797 | 85 | ||
| AK075861.1 | - | 5 - 6 | 771 | 85 | ||
| BC025878.1 | - | 5 - 6 | 741 | 5253854 | 85 | |
| CT010402.1 | - | 4 - 5 | 548 | 85 | ||
| EU886466.1 | - | 5 - 6 | 787 | 85 | ||
| FJ621493.1 | - | 5 - 6 | 785 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sirt3
Genomic Map