Assay Details
Gene Symbol: Cyp2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021282.2 | NP_067257.1 | 4 - 5 | 668 | 111 | |
GenBank mRNA | BC013451.1 | - | 4 - 5 | 668 | 4223797 | 111 |
BC042693.1 | - | 4 - 5 | 647 | 111 | ||
L11650.1 | - | 4 - 5 | 761 | 111 | ||
X62595.1 | - | 4 - 5 | 653 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
