Assay Details
| Gene Symbol: Cdhr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114322.1 | NP_001107794.1 | 10 - 11 | 1257 | 132 | |
| NM_028069.3 | NP_082345.1 | 10 - 11 | 1257 | 132 | ||
| GenBank mRNA | AF462391.1 | - | 10 - 11 | 1246 | 132 | |
| AK007988.1 | - | 10 - 11 | 1246 | 132 | ||
| AK027913.1 | - | 10 - 11 | 1258 | 132 | ||
| BC043336.1 | - | 10 - 11 | 1222 | 5042289 | 132 | |
| BC054469.1 | - | 10 - 11 | 1242 | 4225164 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdhr5
Genomic Map