Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282072.1 | NP_001269001.1 | 10 - 11 | 1585 | 95 | |
| NM_001282073.1 | NP_001269002.1 | 10 - 11 | 1339 | 95 | ||
| NM_001282076.1 | NP_001269005.1 | 11 - 12 | 1542 | 95 | ||
| NM_016874.3 | NP_058570.1 | 9 - 10 | 1415 | 95 | ||
| GenBank mRNA | AF102818.1 | - | 9 - 10 | 1275 | 95 | |
| AK082115.1 | - | 9 - 10 | 1268 | 95 | ||
| AK144333.1 | - | 10 - 11 | 95 | |||
| AK146546.1 | - | 9 - 10 | 1292 | 95 | ||
| AK155836.1 | - | 10 - 11 | 1551 | 95 | ||
| AK158586.1 | - | 10 - 11 | 1224 | 95 | ||
| BC046399.1 | - | 10 - 11 | 1317 | 5063995 | 95 | |
| CD353366.1 | - | 3 - 4 | 351 | 95 | ||
| CF538574.1 | - | 1 - 2 | 117 | 95 | ||
| FJ377318.1 | - | 11 - 12 | 1402 | 95 | ||
| FJ377319.1 | - | 9 - 10 | 1260 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Deaf1
Genomic Map