Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282076.1 | NP_001269005.1 | 12 - 13 | 1786 | 79 | |
| GenBank mRNA | AK082115.1 | - | 10 - 11 | 1512 | 79 | |
| FJ377318.1 | - | 12 - 13 | 1646 | 79 | ||
More Information
Set Membership: |
|

Deaf1
Genomic Map