Assay Details
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177576.1 | NP_001171047.1 | 1 - 2 | 64 | 81 | |
| GenBank mRNA | AK155276.1 | - | 1 - 2 | 58 | 81 | |
| BC037949.1 | - | 1 - 2 | 63 | 5367108 | 81 | |
| BY296753.1 | - | 1 - 2 | 65 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155276.1, BC037949.1, NM_001177576.1 transcripts.
Set Membership: |
|

Slc25a22
Genomic Map