Assay Details
| Gene Symbol: Pidd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022654.1 | NP_073145.1 | 11 - 12 | 2133 | 77 | |
| GenBank mRNA | AF274973.1 | - | 11 - 12 | 2133 | 77 | |
| AK004702.1 | - | 11 - 12 | 2153 | 77 | ||
| AK156863.1 | - | 13 - 14 | 2121 | 77 | ||
| BC032202.1 | - | 11 - 12 | 2010 | 77 | ||
| BC056987.1 | - | 11 - 12 | 2484 | 77 | ||
| BC089373.1 | - | 11 - 12 | 2105 | 77 | ||
| BC099686.1 | - | 11 - 12 | 2471 | 77 | ||
| BC145314.1 | - | 11 - 12 | 2137 | 77 | ||
| BC145315.1 | - | 10 - 11 | 2175 | 77 | ||
| BC145857.1 | - | 11 - 12 | 2098 | 40131160 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pidd1
Genomic Map