Assay Details
| Gene Symbol: Pidd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004702.1 | - | 5 - 6 | 1074 | 78 | |
| BC032202.1 | - | 4 - 5 | 1092 | 78 | ||
| BC099686.1 | - | 4 - 5 | 1469 | 78 | ||
More Information
Set Membership: |
|

Pidd1
Genomic Map