Assay Details
| Gene Symbol: Chst15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029935.5 | NP_084211.2 | 4 - 5 | 1596 | 71 | |
| GenBank mRNA | AB187269.1 | - | 3 - 4 | 1036 | 71 | |
| AF320786.1 | - | 3 - 4 | 1087 | 71 | ||
| AK019474.1 | - | 4 - 5 | 1551 | 71 | ||
| AK031745.1 | - | 2 - 3 | 297 | 71 | ||
| AK043803.1 | - | 2 - 3 | 444 | 71 | ||
| AK082318.1 | - | 4 - 5 | 1662 | 71 | ||
| AK083614.1 | - | 4 - 5 | 1596 | 71 | ||
| AK122324.1 | - | 4 - 5 | 1897 | 71 | ||
| BC031443.1 | - | 4 - 5 | 1553 | 4240286 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chst15
Genomic Map