Assay Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025826.4 | NP_080102.1 | 2 - 3 | 342 | 70 | |
| GenBank mRNA | AK004889.1 | - | 2 - 3 | 243 | 70 | |
| AK076535.1 | - | 2 - 3 | 243 | 70 | ||
| AK131628.1 | - | 2 - 3 | 289 | 70 | ||
| AK152325.1 | - | 2 - 3 | 233 | 70 | ||
| BC011235.1 | - | 2 - 3 | 223 | 71 | ||
| BC035231.1 | - | 2 - 3 | 208 | 70 | ||
| BC044826.1 | - | 2 - 3 | 276 | 69 | ||
| BC054428.1 | - | 2 - 3 | 284 | 5039627 | 70 | |
| BY296718.1 | - | 2 - 3 | 343 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acadsb
Genomic Map