Assay Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025826.4 | NP_080102.1 | 3 - 4 | 452 | 64 | |
| GenBank mRNA | AK004889.1 | - | 3 - 4 | 353 | 64 | |
| AK076535.1 | - | 3 - 4 | 353 | 64 | ||
| AK131628.1 | - | 3 - 4 | 399 | 64 | ||
| AK152325.1 | - | 3 - 4 | 343 | 64 | ||
| BC011235.1 | - | 3 - 4 | 334 | 64 | ||
| BC035231.1 | - | 3 - 4 | 318 | 64 | ||
| BC044826.1 | - | 3 - 4 | 385 | 64 | ||
| BC054428.1 | - | 3 - 4 | 394 | 5039627 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acadsb
Genomic Map