Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700016872.mm.1 |
TC0700001922.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016872.mm.1 |
NM_025826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016872.mm.1 |
NM_025826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016871.mm.1 |
TC0700001922.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016871.mm.1 |
NM_025826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016871.mm.1 |
NM_025826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016868.mm.1 |
TC0700001922.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016868.mm.1 |
NM_025826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016868.mm.1 |
NM_025826.4 |
Refseq |