Assay Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025826.4 | NP_080102.1 | 5 - 6 | 828 | 67 | |
| GenBank mRNA | AK004889.1 | - | 5 - 6 | 729 | 67 | |
| AK076535.1 | - | 5 - 6 | 729 | 67 | ||
| AK131628.1 | - | 5 - 6 | 775 | 67 | ||
| AK152325.1 | - | 5 - 6 | 717 | 67 | ||
| AK163322.1 | - | 1 - 2 | 309 | 67 | ||
| BC011235.1 | - | 5 - 6 | 710 | 67 | ||
| BC035231.1 | - | 5 - 6 | 694 | 67 | ||
| BC044826.1 | - | 5 - 6 | 761 | 67 | ||
| BC054428.1 | - | 5 - 6 | 770 | 5039627 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acadsb
Genomic Map