Assay Details
| Gene Symbol: Pde1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009978.1 | NP_001009978.1 | 11 - 12 | 1211 | 77 | |
| NM_001009979.1 | NP_001009979.1 | 11 - 12 | 1211 | 77 | ||
| NM_001159582.1 | NP_001153054.1 | 13 - 14 | 1713 | 77 | ||
| NM_016744.4 | NP_058024.2 | 12 - 13 | 1508 | 77 | ||
| GenBank mRNA | AF159298.1 | - | 11 - 12 | 1211 | 77 | |
| AK014810.1 | - | 11 - 12 | 1207 | 77 | ||
| AK043647.1 | - | 12 - 13 | 1509 | 77 | ||
| AK050004.1 | - | 13 - 14 | 1575 | 77 | ||
| AY845863.1 | - | 12 - 13 | 1260 | 77 | ||
| AY845864.1 | - | 11 - 12 | 1211 | 77 | ||
| BC090628.1 | - | 12 - 13 | 1476 | 6842675 | 77 | |
| U56649.1 | - | 13 - 14 | 1444 | 77 | ||
More Information
Set Membership: |
|

Pde1a
Genomic Map