Assay Details
| Gene Symbol: Immp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028260.2 | NP_082536.1 | 5 - 6 | 552 | 145 | |
| GenBank mRNA | AK005356.1 | - | 5 - 6 | 553 | 145 | |
| AK009570.1 | - | 5 - 6 | 510 | 145 | ||
| AK010075.1 | - | 5 - 6 | 510 | 145 | ||
| AK011278.1 | - | 5 - 6 | 553 | 145 | ||
| AK011382.1 | - | 5 - 6 | 553 | 145 | ||
| AK011420.1 | - | 5 - 6 | 514 | 145 | ||
| AK011466.1 | - | 5 - 6 | 525 | 145 | ||
| AK011618.1 | - | 5 - 6 | 514 | 145 | ||
| AK011872.1 | - | 5 - 6 | 516 | 145 | ||
| AK012181.1 | - | 505 | 145 | |||
| AK015978.1 | - | 5 - 6 | 523 | 145 | ||
| AK078300.1 | - | 5 - 6 | 539 | 145 | ||
| AK135670.1 | - | 5 - 6 | 512 | 145 | ||
| AK168202.1 | - | 5 - 6 | 501 | 145 | ||
| BC008259.1 | - | 5 - 6 | 511 | 2649406 | 145 | |
| BC081433.1 | - | 4 - 5 | 395 | 6814816 | 145 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Immp1l
Genomic Map