Assay Details
| Gene Symbol: Elp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023876.3 | NP_076365.2 | 3 - 4 | 388 | 100 | |
| GenBank mRNA | AJ276004.1 | - | 3 - 4 | 387 | 100 | |
| AK020742.1 | - | 3 - 4 | 396 | 100 | ||
| AK034040.1 | - | 3 - 4 | 393 | 100 | ||
| AK034529.1 | - | 3 - 4 | 394 | 100 | ||
| AK037104.1 | - | 3 - 4 | 412 | 100 | ||
| AK039447.1 | - | 3 - 4 | 400 | 100 | ||
| AK039758.1 | - | 3 - 4 | 389 | 100 | ||
| AK079069.1 | - | 3 - 4 | 403 | 100 | ||
| AK168964.1 | - | 3 - 4 | 452 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elp4
Genomic Map