Assay Details
| Gene Symbol: Gpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145820.1 | NP_001139292.1 | 14 - 15 | 2199 | 96 | |
| NM_010274.3 | NP_034404.3 | 14 - 15 | 2074 | 96 | ||
| GenBank mRNA | AB045714.1 | - | 1 - 2 | 52 | 96 | |
| AK034353.1 | - | 14 - 15 | 1983 | 96 | ||
| AK144716.1 | - | 14 - 15 | 2200 | 96 | ||
| AK151189.1 | - | 14 - 15 | 1987 | 96 | ||
| AK167152.1 | - | 14 - 15 | 2012 | 96 | ||
| BC021359.1 | - | 14 - 15 | 2168 | 5012142 | 96 | |
| D50430.1 | - | 14 - 15 | 1976 | 96 | ||
| U60987.1 | - | 14 - 15 | 2010 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpd2
Genomic Map