Assay Details
| Gene Symbol: Gpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145820.1 | NP_001139292.1 | 8 - 9 | 1291 | 67 | |
| NM_010274.3 | NP_034404.3 | 8 - 9 | 1166 | 67 | ||
| GenBank mRNA | AK034353.1 | - | 8 - 9 | 1075 | 67 | |
| AK144716.1 | - | 8 - 9 | 1292 | 67 | ||
| AK151189.1 | - | 8 - 9 | 1079 | 67 | ||
| AK167152.1 | - | 8 - 9 | 1104 | 67 | ||
| BC021359.1 | - | 8 - 9 | 1260 | 5012142 | 67 | |
| D50430.1 | - | 8 - 9 | 1068 | 67 | ||
| U60987.1 | - | 8 - 9 | 1102 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpd2
Genomic Map