Assay Details
| Gene Symbol: Lsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271508.1 | NP_001258437.1 | 4 - 5 | 923 | 90 | |
| NM_001271509.1 | NP_001258438.1 | 4 - 5 | 556 | 90 | ||
| NM_001271523.1 | NP_001258452.1 | 4 - 5 | 755 | 90 | ||
| GenBank mRNA | AK075783.1 | - | 4 - 5 | 756 | 90 | |
| BC003796.1 | - | 4 - 5 | 615 | 3488528 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271523.1 transcripts.
Set Membership: |
|

Lsp1
Genomic Map