Assay Details
| Gene Symbol: Xrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011917.2 | NP_036047.2 | 3 - 4 | 379 | 160 | |
| GenBank mRNA | AK004800.1 | - | 3 - 4 | 354 | 160 | |
| AK011711.1 | - | 3 - 4 | 377 | 160 | ||
| AK031247.1 | - | 3 - 4 | 422 | 160 | ||
| AK053643.1 | - | 3 - 4 | 372 | 160 | ||
| AK133654.1 | - | 3 - 4 | 361 | 160 | ||
| AK163899.1 | - | 3 - 4 | 357 | 160 | ||
| AK167019.1 | - | 3 - 4 | 399 | 160 | ||
| AK168037.1 | - | 3 - 4 | 425 | 160 | ||
| BC052395.1 | - | 3 - 4 | 355 | 160 | ||
| BC052823.1 | - | 3 - 4 | 344 | 160 | ||
| BC054743.1 | - | 3 - 4 | 379 | 6400974 | 160 | |
| D38517.1 | - | 3 - 4 | 317 | 160 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xrn2
Genomic Map