Assay Details
| Gene Symbol: Olfr1258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146978.1 | NP_667189.1 | 1 | 545 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149714 | NM_146978.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147532 | NM_146978.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168310 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147532 | NM_146978.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149714 | NM_146978.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P305854 | NM_146978.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415179 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168310 | - | - |
| Whole Mouse Genome G4122A | A_51_P305854 | NM_146978.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113478 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168310 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01150821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01163201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01163206 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01163204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02228911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02206241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02206242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01150812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01150821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01163206 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10473628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473627 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473626 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 600380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2760050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760050 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237353 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237353 | - | - |
| mouse-6 v1.1 (Illumina) | scl4890.1.1_239-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760050 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4890.1.1_239-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760050 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237353 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237353 | - | - |

Olfr1258
Genomic Map